Learn More
Apexbio Technology LLC Eltrombopag Olamine,10mg CAS# 496775-62-3
SDP
Catalog No. 502240616
Encompass_Preferred
Eltrombopag is a new, orally active thrombopoietin-receptor (c-mpl) agonist that stimulates thrombopoiesis. Potential advantages of eltrombopag may include a sustained platelet response and a good tolerability profile. in vitro: Eltrombopag demonstrated a half maximal effective concentration (EC50) of 0.27 uM in murine BAF3 cells transfected with the luciferase reporter gene under direction of the STAT-activated IRF-1 promoter and human TpoR (BAF3/IRF-1/hTpoR) [1]. Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0616 Supplier Apexbio Technology LLC Supplier No. A338710
Eltrombopag is a new, orally active thrombopoietin-receptor (c-mpl) agonist that stimulates thrombopoiesis. Potential advantages of eltrombopag may include a sustained platelet response and a good tolerability profile. in vitro: Eltrombopag demonstrated a half maximal effective concentration (EC50) of 0.27 uM in murine BAF3 cells transfected with the luciferase reporter gene under direction of the STAT-activated IRF-1 promoter and human TpoR (BAF3/IRF-1/hTpoR) [1]. Other sizes are also available. Please inqury us for quote.