Learn More
Medchemexpress LLC Eltrombopag Olamine | 496775-62-3 | C29H36N6O6 | 1 ML
SDP
Catalog No. 5000373010
Encompass_Preferred
Eltrombopag Olamine is an orally active, nonpeptide agonist of the thrombopoietin receptor. It exhibits thrombopoietic activity and has been investigated for its use in treating low blood platelet counts associated with chronic immune thrombocytopenia. Additionally, it is being researched for cardiovascular applications and has demonstrated significant inhibitory effects against multidrug-resistant Staphylococcus aureus. It can also induce apoptosis in hepatocellular carcinoma (HCC) cells.
  • Orally active thrombopoietin receptor nonpeptide agonist
  • Exhibits thrombopoietic activity
  • Highly inhibitory against multidrug-resistant Staphylococcus aureus
  • Induces apoptosis in hepatocellular carcinoma (HCC) cells
  • Activates p-STAT5 and stimulates cell proliferation
  • Increases differentiation of bone marrow CD34+ cells
  • Exhibits antiapoptotic effects in N2C-Tpo cells
Catalog No. 50-003-73010 Supplier Medchemexpress LLC Supplier No. HY15306A10MM/1ML
Eltrombopag Olamine is an orally active, nonpeptide agonist of the thrombopoietin receptor. It exhibits thrombopoietic activity and has been investigated for its use in treating low blood platelet counts associated with chronic immune thrombocytopenia. Additionally, it is being researched for cardiovascular applications and has demonstrated significant inhibitory effects against multidrug-resistant Staphylococcus aureus. It can also induce apoptosis in hepatocellular carcinoma (HCC) cells.
  • Orally active thrombopoietin receptor nonpeptide agonist
  • Exhibits thrombopoietic activity
  • Highly inhibitory against multidrug-resistant Staphylococcus aureus
  • Induces apoptosis in hepatocellular carcinoma (HCC) cells
  • Activates p-STAT5 and stimulates cell proliferation
  • Increases differentiation of bone marrow CD34+ cells
  • Exhibits antiapoptotic effects in N2C-Tpo cells