Learn More
Selleck Chemical LLC Eltrombopag Olamine-5mg
SDP
Catalog No. 502389714
Encompass_Preferred
Eltrombopag Olamine is a member of the biarylhydrazone class, which is a nonpeptide agonist of the thrombopoietin receptor (TpoR).
Catalog No. 50-238-9714 Supplier Selleck Chemical LLC Supplier No. S22295MG
Eltrombopag Olamine is a member of the biarylhydrazone class, which is a nonpeptide agonist of the thrombopoietin receptor (TpoR).