Learn More
TARGETMOL CHEMICALS INC ELUBRIXIN 5MG
SDP
Catalog No. 502581555
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Elubrixin (SB-656933) inhibits neutrophil CD11b upregulation (IC50 of 260.7 nM) and shape change (IC50 of 310.5 nM). Elubrixin can be used for inflammatory diseases such as inflammatory bowel disease and airway inflammation. Elubrixin is a potent selective competitive reversible and orally active CXCR2 antagonist and an IL-8 receptor antagonist. purity: 98%
Catalog No. 50-258-1555 Supplier TARGETMOL CHEMICALS INC Supplier No. T111795MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Elubrixin (SB-656933) inhibits neutrophil CD11b upregulation (IC50 of 260.7 nM) and shape change (IC50 of 310.5 nM). Elubrixin can be used for inflammatory diseases such as inflammatory bowel disease and airway inflammation. Elubrixin is a potent selective competitive reversible and orally active CXCR2 antagonist and an IL-8 receptor antagonist. purity: 98%