Learn More
Pickering Laboratories Inc ELUENT 4X950ML CASE
SDP
Catalog No. NC9200910
Encompass_Preferred
Glyphosate Potassium Phosphate Eluant, case of 4 (950 mL/bottle)
Catalog No. NC9200910 Supplier Pickering Laboratories Inc Supplier No. K200
Glyphosate Potassium Phosphate Eluant, case of 4 (950 mL/bottle)