Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arkray Eluent 80A
SDP

Catalog No. 21377288
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
21-377-288 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Handle with care. Avoid inhalation and contact with skin or eyes. Use in a well-ventilated area. Store away from heat or flames.
Catalog No. 21-377-288 Supplier Arkray Supplier No. 103395
Add to Cart
Add to Cart
Handle with care. Avoid inhalation and contact with skin or eyes. Use in a well-ventilated area. Store away from heat or flames.

Arkray Eluent 80A delivers precise, stable reagent performance for compatible Arkray analyzers. Formulated to support accurate separation and measurement, Eluent 80A helps maintain consistent analytical results and reliable system operation.

  • Designed for Arkray Analyzers: Arkray Eluent 80A is formulated specifically for use in compatible Arkray diagnostic systems, ensuring stable performance and reliable analytical output.
  • Supports Accurate Separation: Provides consistent elution characteristics to maintain precise chromatographic separation within the analyzer.
  • Enhances Measurement Accuracy: Helps ensure accurate quantification and strong reproducibility across multiple test runs.
  • Optimized for System Stability: Contributes to smooth internal fluid flow and stable system pressure, supporting uninterrupted operation.
  • High-Purity Formulation: Manufactured with stringent quality controls to minimize impurities that could impact results or system performance.
  • Consistent Lot Quality: Each batch is tested to ensure consistent chemical composition and performance, reducing variability across testing cycles.
  • Supports High-Throughput Labs: Reliable reagent performance helps laboratories maintain efficient workflow and high test volume capacity.
  • Easy Integration: Designed for quick replacement and minimal preparation time, contributing to faster maintenance and reduced downtime.
  • Improves Analyzer Longevity: Formulated to support optimal internal component function, helping extend the life of compatible Arkray systems.
  • Suitable for Clinical Environments: Ideal for hospitals, reference laboratories, and diagnostic facilities requiring dependable reagent accuracy and stability.

Specifications

CE Marker Yes
Clia Complexity High, Moderate
DoA Calibrators No
Type Reagent
Form Liquid

Optimized for reliable and consistent analytical performance. Compatible with a wide range of Arkray systems for accurate results.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.