Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arkray Eluent 80B
SDP

Catalog No. 21377289
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
21-377-289 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Handle with care. Avoid inhalation and contact with skin or eyes. Use in a well-ventilated area. Store away from heat or flames.
Catalog No. 21-377-289 Supplier Arkray Supplier No. 103396
Add to Cart
Add to Cart
Handle with care. Avoid inhalation and contact with skin or eyes. Use in a well-ventilated area. Store away from heat or flames.

Arkray Eluent 80B ensures precise and consistent chromatography results. Designed for Arkray systems, it provides reliable separation and reproducible performance, making analysis faster and more accurate. Easy to use and ready for immediate application.

  • Reliable Performance: Arkray Eluent 80B delivers precise, reproducible chromatographic results, giving you confidence in every analysis.
  • Optimized for Arkray Systems: Formulated to work seamlessly with Arkray analyzers, ensuring maximum efficiency and accuracy.
  • Ready-to-Use Convenience: Pre-prepared solution saves time and effort, letting your lab focus on results instead of preparation.
  • Consistent Quality: Each batch meets strict quality standards, providing reliable performance run after run.
  • Enhanced Separation: Sharp peaks and improved resolution help produce clear, interpretable chromatograms.
  • Workflow Efficiency: Streamlined setup and minimal maintenance reduce downtime and improve laboratory productivity.
  • Wide Application: Compatible with multiple Arkray assays, making it a versatile choice for diverse testing needs.
  • Stable and Dependable: Maintains stability under standard lab storage conditions, reducing the need for frequent replacement.
  • Reduced Errors: Minimizes variability, ensuring reproducible results across operators and runs.
  • Trusted Brand: Backed by Arkray's legacy of quality and reliability in clinical diagnostics.

Specifications

CE Marker Yes
Clia Complexity High, Moderate
DoA Calibrators Calibration Chip
Type Reagent
Form Liquid

Arkray Eluent 80B is optimized for consistent, reliable chromatography results. Designed for seamless use with Arkray analyzers to improve efficiency and accuracy.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.