Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arkray Eluent 80CV
SDP

Catalog No. 21377290
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
21-377-290 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Handle with care. Avoid inhalation and contact with skin or eyes. Use in a well-ventilated area. Store away from heat or flames.
Catalog No. 21-377-290 Supplier Arkray Supplier No. 103397
Add to Cart
Add to Cart
Handle with care. Avoid inhalation and contact with skin or eyes. Use in a well-ventilated area. Store away from heat or flames.

Arkray Eluent 80CV provides consistent and accurate chromatographic separation. Optimized for Arkray analyzers, it ensures reliable results, reproducibility, and improved workflow efficiency in routine laboratory analyses.

  • Precise Separation: Arkray Eluent 80CV delivers accurate and reproducible chromatographic results, ensuring confidence in every analysis.
  • Optimized for Arkray Systems: Formulated specifically for Arkray analyzers to provide maximum performance and reliability.
  • Ready-to-Use Solution: Pre-prepared eluent saves time and reduces preparation errors, allowing for immediate application in routine workflows.
  • Consistent Quality: Each batch undergoes strict quality control, guaranteeing consistent performance across multiple runs.
  • Enhanced Resolution: Sharp peak definition and improved separation efficiency enhance data quality and interpretation.
  • Workflow Efficiency: Simplifies lab operations with minimal setup and maintenance, reducing downtime and increasing productivity.
  • Wide Application: Suitable for various Arkray assays, making it versatile for multiple analytical needs.
  • Stable and Dependable: Maintains chemical stability under standard laboratory storage conditions, reducing the need for frequent replacement.
  • Minimized Variability: Supports reproducible results across different operators and instruments, ensuring reliability in testing.
  • Trusted Brand: Backed by Arkray's legacy of quality in clinical diagnostics, providing confidence in every use.

Specifications

CE Marker Self-declared
Clia Complexity High, Moderate
DoA Calibrators No
Type Reagent
Form Liquid

Arkray Eluent 80CV is designed for reliable, consistent chromatography results. Optimized for Arkray analyzers, it enhances workflow efficiency and analytical accuracy.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.