Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eluent Solution, for Ion Chromatography, 3 mM Na2CO3/2 mM NaOH, Ricca Chemical
SDP

Catalog No. 282032 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2820-32 1 L
2820-1 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2820-32 Supplier Ricca Chemical Company Supplier No. 282032
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Ion Chromatography

Specifications

Chemical Name or Material Eluent Solution
Color Colorless
Name Note 3mM Na
CAS 7732-18-5, 497-19-8, 1310-73-2
CAS Min % 98.0, 0.03, 0.02
CAS Max % 100.0, 0.03, 0.02
Grade Laboratory
Packaging Natural Poly Bottle
Quantity 1 L
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.