Learn More
Glass Expansion Eluo HF Nebulizer Cleaner for new OpalMist, PolyCon or DuraMist (N0777712)
SDP
Supplier:  Glass Expansion 70ELUOOPD
Encompass_Preferred
Eluo Nebulizer Cleaner for OpalMist and DuraMist Nebulizers. Compatible with Glass Expansion OpalMist and DuraMist nebulizers. Used to clear nebulizer clogs. If the sample capillary becomes blocked with particulates, use the Eluo Nebulizer Cleaning Tool to backflush the blockage. The Eluo is only recommended for Glass Expansion OpalMist and DuraMist nebulizers. For Glass Expansion¿s SeaSpray, MicroMist, Conikal, and Slurry nebulizers, use the Eluo Glass Concentric version (NC9462559).If nebulizer blockages are frequent, consider adding the re-useable Guardian in-line filter (NC0897716). The Guardian in-line filter provides a simple and effective way to eliminate the risk of a blockage, and the filter can be cleaned using a simple attachment for the Eluo (NC0801128). Fluka RBS-25 Concentrate 1L (NC9000117) is an excellent cleaning solution for nebulizers and other sample introduction components.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7353