Learn More
Thermo Scientific™ Elution Buffer for GeneJET Plasmid Midiprep Kit
Shop All Thermo Scientific ProductsDescription
Thermo Scientific GeneJET Elution Buffer is a component of the GeneJET Plasmid Midiprep Kit (Cat. No. K0481) and may be purchased separately. Provided in 60 mL aliquots.
Specifications
Specifications
| Buffer | Elution Buffer |
| For Use With (Application) | Plasmid Prep |
| Product Line | GeneJET |
| Quantity | 60 mL |
| Scale | Midi |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.