Learn More
INNOVAPREP LLC Elution Fluid Can, Tris-buffered solution, Water >99.9%, Tween 20 0.075%, Carbon Dioxide propellant, 125 psi pressure, 50 extractions per can, Non-flammable gas
SDP
Catalog No. NC2032275
Encompass
The InnovaPrep HC08001 Elution Fluid Can is a specially formulated Tris-buffered solution designed for efficient recovery of particles using the InnovaPrep Concentrating Pipette systems. This carbonated elution fluid employs patented Wet Foam Elution™ technology for high recovery rates of microorganisms and particles, maintaining their integrity for various analytical applications. The pressurized can format enhances laboratory workflows with its ready-to-use capability, facilitating rapid and reliable sample concentration.
  • Optimized particle recovery through Wet Foam Elution™ technology
  • Preserves integrity for downstream compatibility
  • Supports up to 50 extractions per can for high throughput
  • Pressurized can ensures safe and immediate use
Catalog No. NC2032275 Supplier INNOVAPREP LLC Supplier No. HC08001
The InnovaPrep HC08001 Elution Fluid Can is a specially formulated Tris-buffered solution designed for efficient recovery of particles using the InnovaPrep Concentrating Pipette systems. This carbonated elution fluid employs patented Wet Foam Elution™ technology for high recovery rates of microorganisms and particles, maintaining their integrity for various analytical applications. The pressurized can format enhances laboratory workflows with its ready-to-use capability, facilitating rapid and reliable sample concentration.
  • Optimized particle recovery through Wet Foam Elution™ technology
  • Preserves integrity for downstream compatibility
  • Supports up to 50 extractions per can for high throughput
  • Pressurized can ensures safe and immediate use