Learn More
INNOVAPREP LLC ELUTION FLUID CAN - PBS
SDP
Catalog No. NC2088239
Encompass
FluidPrep Elution Fluid Can - PBS0.075% Tween 20/PBS. Recommended for classical analytical methods.1 can.Each can provides up to 30 extractions.
This item is not returnable. View return policy
Catalog No. NC2088239 Supplier INNOVAPREP LLC Supplier No. HC08000
Add to Cart
Add to Cart
FluidPrep Elution Fluid Can - PBS0.075% Tween 20/PBS. Recommended for classical analytical methods.1 can.Each can provides up to 30 extractions.