Learn More
Cayman Chemical ElutIon Plate 1ea
Supplier: Cayman Chemical 6019771 EA
Elution Plate has been tested and formulated to work exclusively with Cayman’s Protein A/G Coated Plate Immunoprecipitation Kit (Item No. 601970)