Learn More
Zymo Research Corporation Elution Plate, 2 plates
SDP
Catalog No. 50444079
Encompass_Preferred
Adaptable for use with either Silicon-A plates or Zymo-Spin I-96 filtration plates. Capacity is 90 µl per "V" bottom well.
Catalog No. 50-444-079 Supplier Zymo Research Corporation Supplier No. C2003
Add to Cart
Add to Cart
Adaptable for use with either Silicon-A plates or Zymo-Spin I-96 filtration plates. Capacity is 90 µl per "V" bottom well.