Learn More
TARGETMOL CHEMICALS INC ELVITEGRAVIR 25MG
SDP
Catalog No. 502580881
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Elvitegravir (JTK-303) is a Human Immunodeficiency Virus Integrase Strand Transfer Inhibitor. The mechanism of action of elvitegravir is as an HIV Integrase Inhibitor and Cytochrome P450 2C9 Inducer. purity: 99%
Catalog No. 50-258-0881 Supplier TARGETMOL CHEMICALS INC Supplier No. T233225MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Elvitegravir (JTK-303) is a Human Immunodeficiency Virus Integrase Strand Transfer Inhibitor. The mechanism of action of elvitegravir is as an HIV Integrase Inhibitor and Cytochrome P450 2C9 Inducer. purity: 99%