Learn More
Cayman Chemical Elvitegravir, Purity: ≥98%, CAS: 697761-98-1, 50 mg
SDP
Catalog No. 502979884
Encompass_Preferred
A quinolone antibiotic that inhibits HIV-1 integrase (IC50 = 7.2 nM); has antiviral activity against laboratory strains and clinical isolates of HIV-1 in MAGI cells (EC50s = 0.1-1.26 nM) and HIV-2 in PBMCs (EC50s = 0.3-0.9)
Catalog No. 50-297-9884 Supplier Cayman Chemical Supplier No. 1779850 MG
A quinolone antibiotic that inhibits HIV-1 integrase (IC50 = 7.2 nM); has antiviral activity against laboratory strains and clinical isolates of HIV-1 in MAGI cells (EC50s = 0.1-1.26 nM) and HIV-2 in PBMCs (EC50s = 0.3-0.9)