Learn More
Apexbio Technology LLC Elvitegravir (GS-9137) 697761-98-1 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A40705.1
Encompass_Preferred
Elvitegravir (GS-9137) is an integrase inhibitor targeting HIV-1 which impedes viral DNA integration during the replication cycle of human immunodeficiency virus type 1 (HIV-1) Its mechanism involves inhibition of the integrase enzyme-mediated DNA strand transfer reaction thereby preventing the insertion of viral genetic material into host genomic DNA and subsequently decreasing viral replication Reported inhibitory concentrations (IC50) against HIV-1 integrase range approximately from 7 to 8 nM Besides its application in HIV-1 research Elvitegravir demonstrates inhibitory effects against other retroviral integrases such as murine leukemia virus (MLV) and simian immunodeficiency virus (SIV) making it suitable for broader antiviral research use
Catalog No. 50-000-64051