Learn More
Selleck Chemical LLC Elvitegravir (JTK-303) 50mg 697761-98-1 GS-9137
SDP
Catalog No. 505923
Encompass_Preferred
Elvitegravir (GS-9137, JTK-303) is an HIV integrase inhibitor for HIV-1 IIIB, HIV-2 EHO and HIV-2 ROD with IC50 of 0.7 nM, 2.8 nM and 1.4 nM in cell-free assays, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-592-3 Supplier Selleck Chemical LLC Supplier No. S200150MG
Elvitegravir (GS-9137, JTK-303) is an HIV integrase inhibitor for HIV-1 IIIB, HIV-2 EHO and HIV-2 ROD with IC50 of 0.7 nM, 2.8 nM and 1.4 nM in cell-free assays, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.