Learn More
Abcam Elvitegravir, quinolone HIV integrase inhibitor, 100MG
Supplier:  Abcam AB231510100MG
Encompass_Preferred
MW 447.9 Da, Purity >99%. Elvitegravir is an inhibitor of HIV integrase. It inhibits HIV integrase from HIV 1 IIIB (IC₅₀ = 0.7 nM), HIV 2 EHO (IC₅₀ = 2.8 nM) and HIV-2 ROD (IC₅₀ = 1.1 nM) in cell free assays. Elvitegravir blocks integration of HIV cDNA via inhibition of DNA strand transfer. Active against a range of HIV sub-types and against mulit-drug resistant clinical isolates. Potent (IC₅₀ range = 0.21 - 1.15 nM) HIV antiviral activity in various cell-based assays.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-6147