Learn More
Med Vet International Miltex Winged Dental Basic Kit Elevator, Utility, Series 5, 8in X 3-13/16in X 1-1/8in, 3-083005
Catalog No. 502832148
Encompass_Preferred
The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. This is a Miltex Winged Dental Basic Kit Elevator. Elevators are instruments used in dental extractions or surgical procedures. In dental procedures they may be used to loosen teeth prior to forcep extraction, to remove roots or impacted teeth, when teeth are compromised and susceptible to fracture or when they are malpositioned and cannot be reached with forceps. In surgical procedures they can be used for scraping, elevating or dissecting bones or tissues. They are made from premium-grade surgical stainless steel. Specifications: Utility, Series 5, 8in X 3-13/16in X 1-1/8in, 3-083005
Catalog No. 50-283-2148 Supplier Med Vet International Supplier No. MIL3083005
The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. This is a Miltex Winged Dental Basic Kit Elevator. Elevators are instruments used in dental extractions or surgical procedures. In dental procedures they may be used to loosen teeth prior to forcep extraction, to remove roots or impacted teeth, when teeth are compromised and susceptible to fracture or when they are malpositioned and cannot be reached with forceps. In surgical procedures they can be used for scraping, elevating or dissecting bones or tissues. They are made from premium-grade surgical stainless steel. Specifications: Utility, Series 5, 8in X 3-13/16in X 1-1/8in, 3-083005