Learn More
Medchemexpress LLC HY-114231B 5mg Medchemexpress, ELX-02 (disulfate) CAS:2244622-33-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY114231B5MG
Encompass_Preferred
Medchemexpress, HY-114231B 5mg ELX-02 (disulfate) CAS:2244622-33-9 ELX-02 disulfate (NB-124 disulfate) is an investigational, advanced synthetic eukaryotic ribosome selective glycoside (ERSG). ELX-02 disulfate is being developed as a therapy for genetic diseases caused by nonsense mutations. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9310