Learn More
Medchemexpress LLC Elzovantinib | 2271119-26-5 | 99.9% | 409.42 | 100 MG
SDP
Catalog No. 5000438026
Encompass_Preferred
Elzovantinib (TPX-0022) is an oral-active inhibitor targeting SRC, MET, and c-FMS. It effectively suppresses MET autophosphorylation and downstream signaling pathways such as STAT3, ERK, and AKT in various cell lines. In in-vivo studies, Elzovantinib has demonstrated significant tumor regression and inhibition of tumor growth without causing body weight loss.
  • Oral-active inhibitor
  • Targets SRC, MET, and c-FMS
  • Suppresses MET autophosphorylation and downstream signaling
  • Demonstrates tumor regression in animal models
  • Inhibits tumor growth
  • No body weight loss observed in treated mice
Catalog No. 50-004-38026 Supplier Medchemexpress LLC Supplier No. HY111787100MG
Elzovantinib (TPX-0022) is an oral-active inhibitor targeting SRC, MET, and c-FMS. It effectively suppresses MET autophosphorylation and downstream signaling pathways such as STAT3, ERK, and AKT in various cell lines. In in-vivo studies, Elzovantinib has demonstrated significant tumor regression and inhibition of tumor growth without causing body weight loss.
  • Oral-active inhibitor
  • Targets SRC, MET, and c-FMS
  • Suppresses MET autophosphorylation and downstream signaling
  • Demonstrates tumor regression in animal models
  • Inhibits tumor growth
  • No body weight loss observed in treated mice