Learn More
Medchemexpress LLC EM-163 | 1206480-93-4 | 95.0% | 849.88 | 1 MG
SDP
Catalog No. 5000392802
Encompass_Preferred
EM-163 is a summative BB-Loop analog that can alleviate inflammation and prevent death from toxic shock by targeting the TIR domain of MyD88. It can be utilized in the study of SEB poisoning (Staphylococcal enterotoxin B).
  • Alleviates inflammation.
  • Prevents death from toxic shock by targeting the TIR domain of MyD88.
  • Can be used in the study of SEB poisoning.
  • Inhibits the interaction of the TIR-TIR domain.
  • Inhibits the production of TNF-α, IFN-γ, IL-2 and IL-1b induced by SEB in human mononuclear cells.
  • Effectively inhibits the release of proinflammatory cytokines and delays or avoids death from toxic shock caused by SEB in BALB/c mice.
Catalog No. 50-003-92802 Supplier Medchemexpress LLC Supplier No. HY1630351MG
EM-163 is a summative BB-Loop analog that can alleviate inflammation and prevent death from toxic shock by targeting the TIR domain of MyD88. It can be utilized in the study of SEB poisoning (Staphylococcal enterotoxin B).
  • Alleviates inflammation.
  • Prevents death from toxic shock by targeting the TIR domain of MyD88.
  • Can be used in the study of SEB poisoning.
  • Inhibits the interaction of the TIR-TIR domain.
  • Inhibits the production of TNF-α, IFN-γ, IL-2 and IL-1b induced by SEB in human mononuclear cells.
  • Effectively inhibits the release of proinflammatory cytokines and delays or avoids death from toxic shock caused by SEB in BALB/c mice.