Learn More
Medchemexpress LLC EM-163 | 1206480-93-4 | 95.0% | 849.88 g/mol | C44H60IN5O4 | 10 MG
SDP
Catalog No. 5000288478
Encompass_Preferred
EM-163 is a small-molecule research compound reported to act as an anti-inflammatory agent by targeting the TIR domain of MyD88. It has been used in preclinical studies of toxic shock induced by Staphylococcal enterotoxin B (SEB).
  • Targets the TIR domain of MyD88 to inhibit proinflammatory signaling.
  • Demonstrated activity in models of SEB-induced toxic shock.
  • Available in milligram quantities for laboratory research.
  • Characterized by molecular formula C44H60IN5O4 and molecular weight 849.88 g/mol.
  • Provided with purity specification and safety documentation (SDS and COA) for research use.
Catalog No. 50-002-88478 Supplier Medchemexpress LLC Supplier No. HY16303510MG
EM-163 is a small-molecule research compound reported to act as an anti-inflammatory agent by targeting the TIR domain of MyD88. It has been used in preclinical studies of toxic shock induced by Staphylococcal enterotoxin B (SEB).
  • Targets the TIR domain of MyD88 to inhibit proinflammatory signaling.
  • Demonstrated activity in models of SEB-induced toxic shock.
  • Available in milligram quantities for laboratory research.
  • Characterized by molecular formula C44H60IN5O4 and molecular weight 849.88 g/mol.
  • Provided with purity specification and safety documentation (SDS and COA) for research use.