Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MicroBiologics™ Klebsiella pneumoniae ATCC™ BAA-1144™†
SDP

Catalog No. 23017315 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
6/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-017-315 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-017-315 Supplier MicroBiologics™ Supplier No. 01019K
Add to Cart
Add to Cart
This item is not returnable. View return policy

Microbiologics Klebsiella pneumoniae ATCC™ BAA- 1144™† are a eady-to-use swab that contains a single lyophilized pellet of a single microorganism strain.

Rehydrating fluid is built into the Kwik-Stik unit. These microorganisms are reference stock cultures that have been derived from reference culture collections such as American Type Culture Collection.
  • To use, simply crush the rehydrating ampule, mix, remove swab, and inoculate the agar plate
  • Each canister contains 6 Kwik-Stiks
  • Each shipment includes a Certificate of Analysis, MSDS, and complete instructions

Specifications

ATCC® BAA-1144
Biosafety Level N/A
Certifications/Compliance CE, online certificate of analysis
Color Colorless
Content And Storage 2°C to 8°C
Form Lyophilized Pellet
Media Type Kwik-stik™
Product Type QC Microorganism
For Use With (Application) Culture purpose and QC application
Quantity 6/Pk.
Species Klebsiella pneumoniae
Show More Show Less

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.