Learn More
Andwin Scientific EM PAPER P1196A
SDP
Catalog No. NC9443568
Encompass_Preferred
The test paper and color charts are ideal pH indicators. This product comes with three test fields and nonbleeding, noncontaminating indicator dyes. The test papers measure 6 x 5 mm and are sealed inside a 6 x 85 mm plastic strip.
Catalog No. NC9443568 Supplier Andwin Scientific Supplier No. P11196A
The test paper and color charts are ideal pH indicators. This product comes with three test fields and nonbleeding, noncontaminating indicator dyes. The test papers measure 6 x 5 mm and are sealed inside a 6 x 85 mm plastic strip.