Learn More
AdipoGen EM574 (250 µg)
SDP
Supplier:  AdipoGen AGCN20102C250
Encompass_Preferred
Chemical. CAS: 110480-13-2. Formula: C39H69NO12. MW: 744. Semisynthetic from erythromycin A. Shows strong gastrointestinal motor stimulating (GMS) activity. Motilin receptor agonist. Stimulates smooth muscle cell contraction. Stimulates cholinergic neurons in a non-vagal pathway. Enhances gastrointestinal contractile activity and accelerates gastric emptying. Shows orexigenic activity with affinity for the growth-hormone secretagogue receptor (GHS-R, ghrelin receptor).
Catalog No. 50-205-3458