Learn More
Diagnostic Biosystems EMA Epithelial Membrane Antigen (EMA) E29
Catalog No. 502407551
Encompass_Preferred
This antibody reacts with an antigen of 265‐400 kDa belonging to a heterogeneous group of heavily glycosylated proteins called human milk fat globule proteins. It stains both normal and neoplastic cells. Among normal epithelia, it reacts strongly with mammary epithelium and glandular epithelia but shows a patchy staining with squamous epithelium.\n
Catalog No. 50-240-7551 Supplier Diagnostic Biosystems Supplier No. PDM204
This antibody reacts with an antigen of 265‐400 kDa belonging to a heterogeneous group of heavily glycosylated proteins called human milk fat globule proteins. It stains both normal and neoplastic cells. Among normal epithelia, it reacts strongly with mammary epithelium and glandular epithelia but shows a patchy staining with squamous epithelium.\n