Learn More
Newcomer Supply Epithelial Membrane Antigen (EMA) Control Slides, 10/set
SDP
Supplier:  Newcomer Supply 3285A
Encompass
Newcomer Supply Epithelial Membrane Antigen (EMA) Control Slides are for the positive immunohistochemical staining of EMA, a marker of epithelial cells expressed in a variety of normal and neoplastic epithelia, including mammary and squamous epithelium and sweat glands. EMA is also noted to be expressed in a number of malignant lymphomas. Epithelial Membrane Antigen (EMA) Control Slides contain sections of positive staining kidney and negative staining thyroid.
Catalog No. NC2101946