Learn More
Aviva Systems Biology Corp SEMA4A Antibody - N-terminal region (ARP33269_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33269_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SEMA4A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SEMA4A. Alternate names: RP35, SEMB, SEMAB, CORD10, RP11-54H19.2ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54937