Learn More
Apexbio Technology LLC Emamectin Benzoate 155569-91-8 500mg
SDP
Catalog No. 5000097341
Encompass_Preferred
Emamectin Benzoate (CAS 155569-91-8) is a semi-synthetic derivative targeting insect -aminobutyric acid (GABA) receptors It is designed to interact with these receptors thereby disrupting neurosignaling function Emamectin Benzoate exerts its biological activity primarily through receptor binding that impairs neuronal signaling elevates reactive oxygen species (ROS) production and induces DNA oxidative damage followed by cellular apoptosis In vitro studies have revealed IC50 values typically ranging from low nanomolar to micromolar concentrations depending on insect cell lines and experimental conditions Based on these pharmacological properties Emamectin Benzoate holds research potential in neuroscience toxicology oxidative stress assessments and studies of programmed cell death
Catalog No. 50-000-97341 Supplier Apexbio Technology LLC Supplier No. A4757500
Emamectin Benzoate (CAS 155569-91-8) is a semi-synthetic derivative targeting insect -aminobutyric acid (GABA) receptors It is designed to interact with these receptors thereby disrupting neurosignaling function Emamectin Benzoate exerts its biological activity primarily through receptor binding that impairs neuronal signaling elevates reactive oxygen species (ROS) production and induces DNA oxidative damage followed by cellular apoptosis In vitro studies have revealed IC50 values typically ranging from low nanomolar to micromolar concentrations depending on insect cell lines and experimental conditions Based on these pharmacological properties Emamectin Benzoate holds research potential in neuroscience toxicology oxidative stress assessments and studies of programmed cell death