Learn More
Selleck Chemical LLC Emamectin Benzoate S4423-1g
SDP
Catalog No. 503109327
Encompass_Preferred
Emamectin Benzoate (EMB MK-244) activates gamma-amino butyric acid (GABA) transporter Emamectin Benzoate induces of reactive oxygen species (ROS)-mediated DNA damage and apoptosis
Catalog No. 50-310-9327 Supplier Selleck Chemical LLC Supplier No. S44231G
Emamectin Benzoate (EMB MK-244) activates gamma-amino butyric acid (GABA) transporter Emamectin Benzoate induces of reactive oxygen species (ROS)-mediated DNA damage and apoptosis