Learn More
Medchemexpress LLC Emamectin Benzoate (MK-244) | 155569-91-8 | 98.8% | 100 MG
SDP
Catalog No. 5000407637
Encompass_Preferred
Emamectin Benzoate is an orally active nervous system toxicant that operates by binding to the g-aminobutyric (GABA) receptor in insects. It is a semi-synthetic derivative and exhibits broad-spectrum insecticidal and acaricidal activity. This product induces ROS-mediated DNA damage and cell apoptosis. It is a mixture of natural Emamectin B1a benzoate and Emamectin B1b benzoate, with Emamectin B1a benzoate as the main component.
  • Decreases cell viability and induces apoptosis and DNA damage
  • Increases reactive oxygen species (ROS) generation
  • Inhibits cell viability with an IC50 of 3.72 μM
  • Causes marked induction of oxidative damage in liver tissue
Catalog No. 50-004-07637 Supplier Medchemexpress LLC Supplier No. HYB0837100MG
Emamectin Benzoate is an orally active nervous system toxicant that operates by binding to the g-aminobutyric (GABA) receptor in insects. It is a semi-synthetic derivative and exhibits broad-spectrum insecticidal and acaricidal activity. This product induces ROS-mediated DNA damage and cell apoptosis. It is a mixture of natural Emamectin B1a benzoate and Emamectin B1b benzoate, with Emamectin B1a benzoate as the main component.
  • Decreases cell viability and induces apoptosis and DNA damage
  • Increases reactive oxygen species (ROS) generation
  • Inhibits cell viability with an IC50 of 3.72 μM
  • Causes marked induction of oxidative damage in liver tissue