Learn More
Medchemexpress LLC EMAVUSERTIB 100MG
SDP
Catalog No. NC3450476
Encompass_Preferred
Emavusertib /CA-4948/ is a potent and orally active IRAK4/FLT3 inhibitor. Emavusertib has an IC50 of 57 nM for IRAK4 in a FRET kinase assay. Emavusertib shows anti-tumor activity.For research use only.
Catalog No. NC3450476 Supplier Medchemexpress LLC Supplier No. HY135317100MG
Emavusertib /CA-4948/ is a potent and orally active IRAK4/FLT3 inhibitor. Emavusertib has an IC50 of 57 nM for IRAK4 in a FRET kinase assay. Emavusertib shows anti-tumor activity.For research use only.