Learn More
Cayman Chemical EmavusertIb 50mg
SDP
Catalog No. 502909782
Encompass_Preferred
An IRAK4 inhibitor (IC50 = 31.7 nM); selective for IRAK4 over FLT3, Cdk2, Aurora A kinase, GSK3β, and MUSK at 1 µM; inhibits TLR2-, TLR5-, or IL-1R-mediated IL-6 release in isolated human whole blood (IC50s = 989, 696, and 1,375 nM, respectively); reduces tumor volume in an OCI-Ly3 DLBCL mouse xenograft model from 25-200 mg/kg
Catalog No. 50-290-9782 Supplier Cayman Chemical Supplier No. 3684550 MG
An IRAK4 inhibitor (IC50 = 31.7 nM); selective for IRAK4 over FLT3, Cdk2, Aurora A kinase, GSK3β, and MUSK at 1 µM; inhibits TLR2-, TLR5-, or IL-1R-mediated IL-6 release in isolated human whole blood (IC50s = 989, 696, and 1,375 nM, respectively); reduces tumor volume in an OCI-Ly3 DLBCL mouse xenograft model from 25-200 mg/kg