Learn More
Selleck Chemical LLC Emavusertib CA-4948-5mg
SDP
Supplier:  Selleck Chemical LLC S97795MG
Encompass_Preferred
Emavusertib (CA-4948) is a potent and orally bioavailable inhibitor of interleukin-1 receptor-associated kinase 4/FMS-like Tyrosine Kinase 3 (IRAK4/FLT3) with anti-tumor activity.
Catalog No. 50-238-9741