Learn More
Molecular Devices Molecular Devices Microplate Reader, Fluorescence, Dual Monochromator, 250-850 nm
Catalog No. 5000014667
Encompass_Preferred
The Gemini EM is a dual-monochromator fluorescence microplate reader that provides tunable excitation and emission selection for flexible assay development and sensitive fluorescence detection across common plate formats.
  • Dual monochromators enable wavelength selection from 250-850 nm in 1 nm increments.
  • Supports top and bottom reads for a variety of sample formats.
  • Compatible with 6-384 well microplates for diverse throughput needs.
  • Xenon flash lamp and photomultiplier tube detector deliver high sensitivity.
  • Suitable read modes include endpoint, kinetic, spectrum scan, and well scan.
  • Temperature control from ambient +4°C to 45°C for temperature-sensitive assays.
  • Compact footprint: 34 cm x 42 cm x 42 cm (H x W x D).
Catalog No. 50-000-14667 Supplier Molecular Devices Supplier No. EMG
Add to Cart
Add to Cart
Request a Quote
The Gemini EM is a dual-monochromator fluorescence microplate reader that provides tunable excitation and emission selection for flexible assay development and sensitive fluorescence detection across common plate formats.
  • Dual monochromators enable wavelength selection from 250-850 nm in 1 nm increments.
  • Supports top and bottom reads for a variety of sample formats.
  • Compatible with 6-384 well microplates for diverse throughput needs.
  • Xenon flash lamp and photomultiplier tube detector deliver high sensitivity.
  • Suitable read modes include endpoint, kinetic, spectrum scan, and well scan.
  • Temperature control from ambient +4°C to 45°C for temperature-sensitive assays.
  • Compact footprint: 34 cm x 42 cm x 42 cm (H x W x D).