Learn More
MOLTOX Molecular Toxicology Inc EMB Agar (Levine), 20 mL/plate, 20 plates/sleeve, Store in Refrigerator/Dark
SDP
Catalog No. 50948696
Encompass_Preferred
Eosin Methylene Blue Agar (EMB), Levine is a slightly selective and differential plating medium for the isolation of gram-negative bacteria. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-696 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S87
Eosin Methylene Blue Agar (EMB), Levine is a slightly selective and differential plating medium for the isolation of gram-negative bacteria. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.