Learn More
Alkali Scientific EMB Agar (Levine), 20 mL, 100 × 15 mm Plate, 10/Pk
SDP
Catalog No. 5000661996
Encompass_Preferred
EMB Agar (Levine), 20 mL, 100 × 15 mm Plate, 10/Pk is ideal for the cultivation of enteric bacteria. This medium contains bile salts, which inhibit the growth of non-enteric organisms, while providing ideal conditions for the growth of E. coli and other coliforms. The 20mL plates come pre-poured and ready to use for microbiological testing.
Catalog No. 50-006-61996 Supplier Alkali Scientific Supplier No. EMBL00
EMB Agar (Levine), 20 mL, 100 × 15 mm Plate, 10/Pk is ideal for the cultivation of enteric bacteria. This medium contains bile salts, which inhibit the growth of non-enteric organisms, while providing ideal conditions for the growth of E. coli and other coliforms. The 20mL plates come pre-poured and ready to use for microbiological testing.