Learn More
Teknova EMB Agar Plates. 100mm Plates, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502388153
Encompass
1.00% Peptone, 0.20% Dipotassium Phosphate,0.04% Eosin Y Disodium Salt, 1.50% Agar,65g/mL Methylene Blue Hydrate, 2.00% Lactose
Catalog No. 50-238-8153 Supplier Teknova Supplier No. E0070
1.00% Peptone, 0.20% Dipotassium Phosphate,0.04% Eosin Y Disodium Salt, 1.50% Agar,65g/mL Methylene Blue Hydrate, 2.00% Lactose