Learn More
Electron Microscopy Sciences Embed 812 Embedding Kit with BDMA
SDP
Catalog No. 501930502
Encompass_Preferred
Electron Microscopy Sciences' replacement for the discontinued Epon-812. Produces the same results in terms of preservation, handling, sectioning, and staining as the old Epon-812. Kit consists of 450 ml EMbed-812, 450 ml DDSA; Specially Distilled, 450 ml NMA, and 50 ml BDMA.
Catalog No. 50-193-0502 Supplier Electron Microscopy Sciences Supplier No. 14121
Electron Microscopy Sciences' replacement for the discontinued Epon-812. Produces the same results in terms of preservation, handling, sectioning, and staining as the old Epon-812. Kit consists of 450 ml EMbed-812, 450 ml DDSA; Specially Distilled, 450 ml NMA, and 50 ml BDMA.