Learn More
Electron Microscopy Sciences EMbed-812 kit, 1 kit
SDP
Catalog No. 50980391
Encompass_Preferred
Replacement for the discontinued Epon-812. Produces the same results in terms of preservation, handling, sectioning, and staining as the old Epon-812. Kit consists of 450 ml EMbed-812, 450 ml DDSA; Specially Distilled, 450 ml NMA, and *50 ml DMP-30. *Please note that wherever DMP-30 is listed as the accelerator in our embedding media kits you have the option to use BDMA in place of it. BDMA is less viscous, has a longer shelf life, and offers better penetration (it should be noted, however, that in order to achieve the optimum results when using BDMA in place of DMP-30 you must vary the amount. DMP-30: 1.5-2%; BDMA: 2.5-3%).
Catalog No. 50-980-391 Supplier Electron Microscopy Sciences Supplier No. 14120
Replacement for the discontinued Epon-812. Produces the same results in terms of preservation, handling, sectioning, and staining as the old Epon-812. Kit consists of 450 ml EMbed-812, 450 ml DDSA; Specially Distilled, 450 ml NMA, and *50 ml DMP-30. *Please note that wherever DMP-30 is listed as the accelerator in our embedding media kits you have the option to use BDMA in place of it. BDMA is less viscous, has a longer shelf life, and offers better penetration (it should be noted, however, that in order to achieve the optimum results when using BDMA in place of DMP-30 you must vary the amount. DMP-30: 1.5-2%; BDMA: 2.5-3%).