Learn More
Electron Microscopy Sciences Embed 812 Resin 450 ML
SDP
Catalog No. 50980447
Encompass_Preferred
EMbed 812 has the same characteristics as Epon 812, physically, and provides the same results in preservation, handling, curing and sectioning. EMbed 812 may cause etching on selected plastics. All formulas for mixing and using Epon 812 can be applied to EMbed 812, e.g. Luft's procedure, Mollenhauer's and EPON-ARALDITE mixtures.
Catalog No. 50-980-447 Supplier Electron Microscopy Sciences Supplier No. 14901
EMbed 812 has the same characteristics as Epon 812, physically, and provides the same results in preservation, handling, curing and sectioning. EMbed 812 may cause etching on selected plastics. All formulas for mixing and using Epon 812 can be applied to EMbed 812, e.g. Luft's procedure, Mollenhauer's and EPON-ARALDITE mixtures.