Learn More
Electron Microscopy Sciences Embedding Cassettes in QuickLoad™ Sleeves, Gray
SDP
Catalog No. 501930748
Encompass_Preferred
Molded from a special high density acetal polymer, they keep specimens safely submerged and are totally resistant to the chemical action of solvents used in histology laboratories. The efficient flow-through slots maximize fluid exchange and ensure proper reagent drainage.
Color: Gray
This item is not returnable. View return policy
Catalog No. 50-193-0748 Supplier Electron Microscopy Sciences Supplier No. 7007904SL
Molded from a special high density acetal polymer, they keep specimens safely submerged and are totally resistant to the chemical action of solvents used in histology laboratories. The efficient flow-through slots maximize fluid exchange and ensure proper reagent drainage.
Color: Gray