Learn More
Electron Microscopy Sciences Embedding Block 1, 19x19x19 mm
SDP
Catalog No. 5025725
Encompass_Preferred
These embedding blocks are precisely machined from hard phenolic resin. The faces of the embedding blocks are cut very deeply providing a firm hold of the embedded specimen. Suitable for cellodin or paraffin sections.
Size: 19 x 19 x 19 mm
Catalog No. 50-257-25 Supplier Electron Microscopy Sciences Supplier No. 6236301
These embedding blocks are precisely machined from hard phenolic resin. The faces of the embedding blocks are cut very deeply providing a firm hold of the embedded specimen. Suitable for cellodin or paraffin sections.
Size: 19 x 19 x 19 mm