Learn More
Electron Microscopy Sciences Embedding Block 3, 38 x 38 x 25 mm
SDP
Catalog No. 5025724
Encompass_Preferred
These embedding blocks are precisely machined from hard phenolic resin. The faces of the embedding blocks are cut very deeply providing a firm hold of the embedded specimen. Suitable for cellodin or paraffin sections.
Size: 38 x 38 x 25 mm
Catalog No. 50-257-24 Supplier Electron Microscopy Sciences Supplier No. 6236303
These embedding blocks are precisely machined from hard phenolic resin. The faces of the embedding blocks are cut very deeply providing a firm hold of the embedded specimen. Suitable for cellodin or paraffin sections.
Size: 38 x 38 x 25 mm