Learn More
Electron Microscopy Sciences Embedding Block 4, 51 x 51 x 19 mm
SDP
Catalog No. 5025727
Encompass_Preferred
These embedding blocks are precisely machined from hard phenolic resin. The faces of the embedding blocks are cut very deeply providing a firm hold of the embedded specimen. Suitable for cellodin or paraffin sections.
Size: 51 x 51 x 19 mm
Catalog No. 50-257-27 Supplier Electron Microscopy Sciences Supplier No. 6236304
These embedding blocks are precisely machined from hard phenolic resin. The faces of the embedding blocks are cut very deeply providing a firm hold of the embedded specimen. Suitable for cellodin or paraffin sections.
Size: 51 x 51 x 19 mm