Learn More
Electron Microscopy Sciences Embedding Capsule Holder
SDP
Supplier:  Electron Microscopy Sciences 7002204
Encompass_Preferred
Fabricated form polypropylene, a direct substitution for the BEEM capsule holders. 15 positions. Fits embedding capsule size 00' (8.5 mm diameter). Sturdy, long lasting, and stackabe.
Catalog No. 50-257-30