Learn More
Electron Microscopy Sciences Embedding Capsule Substitute
SDP
Catalog No. 5029267
Encompass_Preferred
This 24 cavity mold produces a block similar to the embedding capsule block but with the following advantages: * Resin polymerizes better in silicone rubber than in polypropylene or polyethylene - the rubber heats more evenly and retains the heat better than in plastic. It is easier to see inside the well and to remove block from well. Finished blocks have finer tips.
Catalog No. 50-292-67 Supplier Electron Microscopy Sciences Supplier No. 70165
This 24 cavity mold produces a block similar to the embedding capsule block but with the following advantages: * Resin polymerizes better in silicone rubber than in polypropylene or polyethylene - the rubber heats more evenly and retains the heat better than in plastic. It is easier to see inside the well and to remove block from well. Finished blocks have finer tips.